Would you like to log in again?
SP Wilmad-LabGlass Two-Piece Vacuum Traps with Hose Connection, SP Industries
Supplier: SP Industries
LG-11026-104 LG-11026-304 LG-11026-102 LG-11026-100 LG-11026-306 LG-11026-308 LG-11026-302 LG-11026-106 LG-11026-300 LG-11026-310 LG-11026-110
14203-346EA 255.02 USD InStock
14203-346 14202-922 14210-388 14209-112 14202-726 14207-172 14207-100 14207-142 14208-024 14208-310 14209-962
SP Wilmad-LabGlass Two-Piece Vacuum Traps with Hose Connection, SP Industries
Vacuum Traps
These borosilicate glass vacuum traps have a Standard Taper joint to ease disassembly and allow cleaning/draining
The sidearm 3/8" hose connectors are 180° apart with a bent top stem and provide an easy connection when using rubber tubing. See LG-11020 for body only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty