Would you like to log in again?
SP Wilmad-LabGlass Volumetric Flasks with Plastic Stopper, Class A, SP Industries
Supplier: SP Industries
LG-8116-108 LG-8116-116 LG-8116-122 LG-8116-112 LG-8116-128 LG-8116-126 LG-8116-132 LG-8116-118 LG-8116-106 LG-8116-124 LG-8116-120 LG-8116-110
14210-620EA 194.05 USD InStock
14210-620 14211-012 14209-442 14202-050 14205-558 14202-782 14204-590 14207-000 14207-484 14210-678 14203-310 14208-774
SP Wilmad-LabGlass Volumetric Flasks with Plastic Stopper, Class A, SP Industries
Flasks Volumetric Flasks
These borosilicate glass volumetric flasks are provided with a plastic closed-bottom stopper which fits the Standard Taper ground zone
Stopper design prevents the neck from breaking if flask is tipped over. Calibrated "to contain" (TC) at 20°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty