Would you like to log in again?
SP Wilmad-LabGlass Distilling Heads with Vacuum Port, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LG-6115-126 ML-870-702 LG-6115-100
14209-102EA 660.61 USD InStock
14209-102 14205-244 14208-972
SP Wilmad-LabGlass Distilling Heads with Vacuum Port, SP Industries
Distillation Components Distillation Heads
These distilling heads have a one piece design with the condenser angled at 75° and a vacuum port on product side which can be left open for atmospheric distillations.
  • Borosilicate glass
A Standard Taper 10/30 outer joint is provided for a 75mm immersion thermometer. See thermometer adapters (LG-1453) for use with models ML-870.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty