Would you like to log in again?
SP Wilmad-LabGlass Filtration Apparatus, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BP-1752-001 BP-1781-090 BP-1752-091 BP-1755-000 BP-1763-002 BP-1751-000 BP-1763-000 BP-1763-004 BP-1753-090 BP-1755-090 BP-1753-000
14211-340EA 48.88 USD InStock
14211-340 14204-302 14209-980 14007-308 14207-704 14205-502 14209-150 14210-240 14201-906 14203-068 14007-310
SP Wilmad-LabGlass Filtration Apparatus, SP Industries
Filters Membrane Filter Units
These filter assemblies for the FILTERWARE™ filtration system consist of an all-borosilicate glass funnel, fritted glass support base, aluminum clamp, and silicone stopper.
Ordering information: Available from 47 mm or 90 mm filter assembly sizes. Individual components are offered in various sizes for a customized unit. Designed for use with (14209-536) filter flasks, which are sold separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty