Would you like to log in again?
SP Wilmad-LabGlass Precision Jacketed Glass Sleeve Glass Stirrer Bearing, Heavy Duty, SP Industries
Supplier: SP Industries
LG-9427-140
14209-332EA 519.96 USD InStock
14209-332
SP Wilmad-LabGlass Precision Jacketed Glass Sleeve Glass Stirrer Bearing, Heavy Duty, SP Industries
Stirrers Stirring Tools
Designed for stirring reaction mixtures with increased viscosity, these heavy-duty glass sleeve bearings are precision bore, have 19 mm I.D., and are jacketed with hose connectors for cooling.
Lower end of bearing has a Standard Taper inner joint. Use with glass rod stirrer shaft (LG-9504) and PTFE blade (LG-9516T).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty