Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Wilmad-LabGlass Polished Glass Stirrer Shafts, SP Industries
Supplier: SP Industries
LG-9501-104 LG-9501-100 LG-9501-102
14202-598EA 182.2 USD InStock
14202-598 14210-772 14210-250
SP Wilmad-LabGlass Polished Glass Stirrer Shafts, SP Industries
Stirrers Stirring Rods
Precision ground and polished, these borosilicate glass stirrer shafts have a 10mm O.D
For use with LG-9475T bearing with PTFE bearing surface, or other bearings with a PTFE interface with the same bore. Features a side button at bottom for attachment of any of the stirrer/stirring blades (LG-9505 through LG-9511T).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty