Would you like to log in again?
SP Wilmad-LabGlass Round Bottom Vertical/Angled Two Neck Flasks, SP Industries
Supplier: SP Industries
LG-7291-246 LG-7291-254 LG-7291-278 LG-7291-256 LG-7291-234
14209-156EA 477.6 USD InStock
14209-156 14210-474 14203-030 14202-804 14204-056
SP Wilmad-LabGlass Round Bottom Vertical/Angled Two Neck Flasks, SP Industries
Flasks Round Bottom Flasks
These flasks have a center neck that is vertical and a side neck that is at a 20° angle to the center.
  • Borosilicate glass
Both necks have full length outer Standard Taper joints.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty