Would you like to log in again?
SP Wilmad-LabGlass Flash Chromatography Reservoirs, SP Industries
Supplier: SP Industries
LG-4525-100 LG-4525-102
14210-432EA 237.68 USD InStock
14210-432 14210-628
SP Wilmad-LabGlass Flash Chromatography Reservoirs, SP Industries
Reagent Reservoirs
These reservoirs are used when larger volumes are required to elute samples from flash chromatography column.
  • Borosilicate glass
  • For containment of mobile phase
  • Connects flow controller and column
Upper outer joint connects to flow controller while lower inner joint connects with column.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty