Would you like to log in again?
Septihol® Isopropyl Alcohol Solution, STERIS®
Supplier: Steris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Septihol®
1S3508 1S3596 1S35Y4 625477 625408 625408 625405 625401 624808 625477 1S35Y4
89094-690CS 667.21 USD InStock
CA14211-728 89094-690 10736-414 21899-859 76245-118 14227-718 CA14227-718 CA21899-859 14211-728 14223-886 CA14223-886
Septihol® Isopropyl Alcohol Solution, STERIS®
Cleaning Agents Disinfectants
Septihol® solution is a quick-evaporating 70% (v/v) USP isopropyl alcohol/30% (v/v) USP purified water blend that has been filtered to 0.22 µm. Ideal for the removal of process residues and for routine glove decontamination.
  • Ready-to-use solution
  • Evaporates quickly
  • Formulated without stabilizers or denaturants
  • Used in pharmaceutical, biotechnology and medical device manufacturing areas
Ideal for the removal of process residues from equipment, surfaces, gloves, hoods, countertops and laminar flow curtains.
Sterile version is gamma irradiated and each lot USP sterility tested.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty