Would you like to log in again?
Leakbuster™ Specimen Containers
Supplier: Starplex Scientific
LeakBuster™
B902L B902-1W WB902-1WNL B1202-1W-COS B602-1YNL B1202-1YN B1202-1W-CO B1202-VOS B902-1O B602-VO B1252-10 B1202-1YNL WB902-1YN WB902-1O B602-1O B902-1W B1202-1O-OR B1202-VO B1202-1O B1202-1W-CO V9740 B602-1YN B1202-1YN B1202-10-0R B952-1O B202-VO B902-1O B602-1O B1202-1W-COS B1202-1O B902-10-20 B902-1YN B900-1W-CO B1202-VO B952-10 B602-1YNL B902-VO B902L B602L B202-1W-CO WB902-1O B602L B900-1O-CO WB902-1YN B952-LFS B902-1YNL B902-1YN B1252-1O
10811-010CS 152.55 USD InStock
CA73240-116 CA11006-280 CA73240-038 CA73240-132L CA68810-007 CA10799-212 CA68810-008 CA53594-812 CA73250-004 10811-010 14211-744 CA73240-110 CA55500-046 CA73240-098 CA55500-022 CA55500-023 CA73240-092 10811-012 CA73240-150 55500-156 CA73240-094 CA10798-884 53594-812 CA68810-010 14216-254 CA73240-106 14216-256 14211-752 14216-258 14211-730 14211-750 CA73270-064 14211-734 CA53594-824 CA55500-156 14211-732 CA73240-120 14211-738 14216-250 CA55500-053 CA55500-031 14216-252 14211-736 CA10798-955 CA10798-954 CA53594-810 CA73240-109L CA73240-028L
Leakbuster™ Specimen Containers
Sample Containers General Purpose Containers
This premium quality line of specimen containers consists of 60 ml, 90 ml, wide 90 ml, and 120 ml sizes for leakproof transportation.
Containers are ideal for use in harsh transport conditions or in pneumatic tube systems. All vials are manufactured from clear, flexible, crack-resistant polypropylene. The streamlined straight-sided vials are stable, stackable, and save space in storage. There are graduation markings on three sides. All sterile containers have a tab on the label to ensure the sterility integrity of the container. Midstream kits consist of a LeakBuster™ container and a castile soap wipe. Kit 14211-750 comes with a 90 ml container and an affixed funnel. Kit 14211-752 comes with a 120 ml container.
All caps are designed with a full circle thread, which prevents the cap from cross threading and leaking. The O-ring cap is designed to grip the top and sides of the vial. The vial fits into the groove of the cap for additional contact, and a better seal. The lined cap is manufactured with a polypulp liner coated with polyethylene. Due to the resilience of the lining material, it provides extra protection against leaks. QuickTurn is a cap and vial combination that has a double thread design, which allows the cap to be securely affixed with only a quarter turn.
Each polypropylene container is lot numbered and features clear graduations. The wide 90 and 120 ml containers feature a QuickTurn™ cap closure for easy application. Sterile containers have an orange cap and tamper tab patient label. The patient identification labels meet current JCAHO standards. Nonsterile containers have a yellow cap and a non-tabbed patient label. The standard 90mL container is also available in an opaque polypropylene for discrete transport. A space saver dispenser box of twenty 90 ml containers is convenient for laboratories and doctor's offices. Ideal for operating room use, both the 90 and 120 ml containers are packaged in a sterile, clear plastic peel pouch.
Ordering information: Containers 14216-256 are packaged in dispenser boxes of 20 containers; 20 boxes per case. Containers 14216-258 are individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty