Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Starswabs II® Specimen Collection and Transport Systems
Supplier: Starplex Scientific
Starswabs II®
SP130-W SP130D SP130L SP130-FW S09 SP130D SP132D SP132-W SP130L-W SP132 S09 SP130L-W SP131D SP131D S08 SP130L SP130-W SP132-W SP130X SP130L-D SP132D SP131X SP130L-D SP131-W SP132 SP130X SP131X SP131-W
14211-760CS 722.16 USD InStock
14211-760 14211-764 14211-762 CA66410-106L CA73270-010 14211-768 14211-766 CA73270-052 CA66410-101 CA66410-022 CA66410-028 CA66410-026 66410-032 CA73270-005 14211-774 CA73270-021 14211-756 14211-754 CA66410-105L CA73270-003 14211-758 CA66410-057 66410-026 66410-028 CA66410-103L CA66410-032 CA66410-051 66410-022
Starswabs II® Specimen Collection and Transport Systems
Swabs Transport Swabs
Sterile, ready to use units for the collection and transport of bacteriological specimens
Both Amies provide excellent environments for the survival of anaerobes through the complete immersion of the swabs into the gel and by the presence of the reducing agent sodium thioglycollate. Shelf life is 24 months. Amies Clear is a modified Amies gel that provides excellent survival for a wide range of bacteria and is compatible with gram stain. Amies Charcoal enhances the survival of fastidious organisms by neutralizing any toxins that may be present in the specimen.
Modified Liquid Stuart's provide excellent recovery in routine bacteriology and are compatible with gram stain and many rapid screening methods. Shelf life is 18 months.
Swab length is 15.2cm (6"). Color-coded cap with protective overskirt. Cap rings ensure containment of biohazardous material. Tube label shows code, lot number, and expiration date. Thick tube wall protects medium from dehydration. Spiral media chamber keeps sample immersed and protected during transport. Round-bottom tube for easy racking. Seamless molding ensures tube strength and integrity. Rayon swab tip is inert, non-toxic, and permits good sample retrieval and absorption.
Ordering information: Swabs are packed in bags that create a barrier environment to prevent premature medium dehydration. Bags feature resealable closures and are metalized to protect medium from harmful UV rays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty