Would you like to log in again?
Starswab® Anaerobic Transport System, Starplex®
Supplier: Starplex Scientific
Starswab®
S120-D S120-D
14211-770CS 546.41 USD InStock
14211-770 CA53594-821
Starswab® Anaerobic Transport System, Starplex®
Swabs Transport Swabs
Maintains the viability of a wide variety of microorganisms during transportation for up to 48 hours
Self-contained, sterile, ready-to-use system consists of a glass tube containing semi-solid, pre-reduced transport medium, and two rayon, scored swabs. Features an indicator that will turn lavender if the anaerobic condition is compromised. Pierceable, latex-free septum. Screw cap with rubber center allows fluids to be injected through with needle and syringe. Tube label shows code, lot number, and expiration date. Shelf life is 18 months. Swab length: 15.2 cm (6").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty