Would you like to log in again?
SP Wilmad-LabGlass Serological Color Coded Class B Pipettes, SP Industries
Supplier: SP Industries
LG-9335-120 LG-9335-116 LG-9335-112 LG-9335-110 LG-9335-118
14212-244EA 20.66 USD InStock
14212-244 14212-246 14212-502 14211-580 14212-506
SP Wilmad-LabGlass Serological Color Coded Class B Pipettes, SP Industries
Pipettes Serological Pipettes
Calibrated 'to deliver' (TD) at 20°C, these borosilicate glass pipettes have a permanently colored amber scale with the zero line at the top
  • Reference: ASTM E 1044, Lab Grade
The small amount remaining in the tip after completion of free delivery is blown out and added to the main delivery volume (ground band indicator at top). The 5,10, and 25 ml pipettes have openings of reduced diameter.
Caution: NOTE: Do not pipette by mouth. A mechanical device is suggested.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty