Would you like to log in again?
SP Wilmad-LabGlass Polypropylene Reservoir Storage Cap, SP Industries
Supplier: SP Industries
BP-1909-000
14212-422EA 33.45 USD InStock
14212-422
SP Wilmad-LabGlass Polypropylene Reservoir Storage Cap, SP Industries
Closures Safety Caps and Safety Waste Caps
This resevoir storage cap is red, made from polypropylene, and features a 45 mm thread and PTFE liner.
Fits all CHROMWARE® HPLC reservoirs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty