Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Wilmad-LabGlass PTFE Stirrer Blades, SP Industries
Supplier: SP Industries
LG-9510T-100 LG-9510T-104 LG-9510T-106 LG-9510T-102 LG-9510T-110 LG-9510T-108
14211-576EA 19.95 USD InStock
14211-576 14212-356 14212-498 14212-448 14202-882 14212-428
SP Wilmad-LabGlass PTFE Stirrer Blades, SP Industries
Stirrers Stirring Blades
These PTFE stirrer blades are used with 10 mm outer diameter shafts that have a side button for attachment of blades.
  • 1/8" thick blade
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty