Would you like to log in again?
SP Wilmad-LabGlass Volumetric Flasks with Penny Head Stoppers, Class A, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LG-8110-122 LG-8110-124 LG-8110-134 LG-8110-130 LG-8110-126 LG-8110-104 LG-8110-112 LG-8110-114 LG-8110-118 LG-8110-106 LG-8110-108 LG-8110-120 LG-8110-128 LG-8110-110 LG-8110-132 LG-8110-116 LG-8110-100 LG-8110-102
14201-988EA 41.19 USD InStock
14201-988 14208-638 14209-866 14205-422 14205-342 14209-786 14210-698 14208-062 14209-140 14202-832 14208-120 14206-138 14212-788 14208-014 14211-182 14202-394 14202-076 14202-098
SP Wilmad-LabGlass Volumetric Flasks with Penny Head Stoppers, Class A, SP Industries
Flasks Volumetric Flasks
These borosilicate glass volumetric flask sizes comply with requirements for Type 1, Class A "Unserialized flasks"
Reference: ASTM E288 and Federal Specifications NNN-F-289.
Note: LG-8110-100 and LG-8110-134 are not volumetric shaped.
They are supplied with a standard taper penny head glass stopper. Calibrated "to contain" at 20°C.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty