Would you like to log in again?
MORTAR HARD PORCELAIN
Supplier: VERDER SCIENTIFIC INC.
Hard Porcelain Mortar^ For use with the RM 100 Mortar Grinder. In most cases, grinding sets of hard porcelain are adequate for use on soft, medium-hard or pasty materials. Composition is approximately 71% SiO2, 24% Al2O3. Degree of hardness is 7 Mohs, and the density is approximately 2400kg/m3.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MORTAR HARD PORCELAIN 14213-034 024600016 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty