Would you like to log in again?
WHEATON® ABC Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WHEATON®
W225338-0801 W225338-08 W225333-03 W225330-0107 W225332-0203 W225334A-0401 W225336-06 W225334A-04 W225333-0301 W225330-0101 W225332-0201 W225330-0103 W225332-0207 W225330-0105 W225336-0601 W225332-02 W225338-0804 W225332-0205 W225330-01 W225332-0204 W225330-0104
89235-510CS 428.48 USD InStock
89235-510 89235-512 14213-318 14213-338 14213-316 14213-326 14213-336 14213-314 14213-324 89232-028 14213-334 14213-322 89232-026 14213-300 14213-332 89235-508 14213-320 89232-024 89235-506 89232-022 89232-020
WHEATON® ABC Screw Caps, DWK Life Sciences
Closures Chromatography Vial Closures
These closures for ABC Vials feature Wheaton Super Septa and low extractable septa.
  • Manufactured from polypropylene
  • 100/pack, 10 packs/case
Available in natural, black, red, blue, yellow or green polypropylene
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty