Would you like to log in again?
COVER 0.5 L F. PLASTIC RCPTCL
Supplier: VERDER SCIENTIFIC INC.
Plastic Lid for 0.5L Plastic Vessel^ For use in the GM 200 Laboratory Knife Mill. This cover prevents the sample material from escaping the vessel during the milling process.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVER 0.5 L F. PLASTIC RCPTCL 14214-610 031070310 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty