Would you like to log in again?
Cal-Rite Decalcifing Solution, Epredia
Supplier: Epredia
5501 5501
14214-926EA 296.87 USD InStock
CA14214-926 14214-926
Cal-Rite Decalcifing Solution, Epredia
Stains Microscopy and staining reagents
The Epredia™ Cal-Rite Decalcifing/Fixative Solution is a mixature of formic acid and formaldehyde which permits simultaneous decalcification and fixation.
  • Eliminates poor hematoxylin and eosin staining caused by over decalcification and /or exposure to harsh mineral acids.
  • Conventional iron staining is not hampered by Cal-Rite.
  • Prolonged exposure to Cal-Rite does not distort nuclear chromatin staining
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty