Would you like to log in again?
Alchohol Wipes, Techspray
Supplier: ITW Chemtronics
1610-50PK
14216-032CS 621 USD InStock
14216-032
Alchohol Wipes, Techspray
Wipes Disposable Wipes and Towels Disinfectant Wipes
All-purpose cleaning wipes presaturated with pure, anhydrous isopropyl alcohol.
  • Safe on plastics
  • Non-corrosive
  • Rapidly evaporating
  • Zero residue
  • Non-ozone depleting
Ideal for tape head cleaning, removal of fluxes, light oils, polar soils, and white mineral residue.
Certifications: EPA SNAP Approved.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty