Would you like to log in again?
FLUX REMOVER ECOLINE 10OZSPRAY
Supplier: ITW-CHEMTRONICS,INC
Ecoline Flux Remover^ Dissolves type R, RMA, SA, and most no-clean fluxes. Can also be used to remove light oils, silicones, waxes, greases, and similar contaminants. Safe on most plastics. Non-ozone depleting. Zero residue, and rapidly evaporating. 284g aerosol.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLUX REMOVER ECOLINE 10OZSPRAY 14216-037 1621-10S Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty