Would you like to log in again?
Envi-Ro-Tech™ 1671 Dusters, Techspray™
Supplier: Techspray
Envi-Ro-Tech™
1671-15S 1671-10S
14216-056CS 522.05 USD InStock
14216-056 58542-430
Envi-Ro-Tech™ 1671 Dusters, Techspray™
Sprays
Exceptionally pure, moisture-free inert gas removes particulates without using harmful solvents.
  • Nonabrasive
  • Contain no CFCs
Nonflammable dusters leave no residue and are quad-filtered.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty