Would you like to log in again?
Glass Dropping Bottles, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
W211755 W211735 W211754 W211734
14216-232CS 153.85 USD InStock
14216-232 14216-234 14216-236 14216-230
Glass Dropping Bottles, Wheaton®, DWK Life Sciences
Bottles Dropping and Dispensing Bottles
With glass hood stopper.
Channel in stopper allows drop-by-drop delivery of contents when bottle is tipped. A partial turn of the stopper seals bottle. Available in clear and amber glass. USD Type III Glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty