Would you like to log in again?
Electronic Balance Calibration Weights, ANSI/ASTM Class 2 Weight, Troemner
Supplier: Troemner
8253 8232 8228 8200T 8206T 8200 8242 8254 8236 8252T 8234W 8228W 8244W 8224 8216 8202 8206W 8200W 8216W 8214W 8244 8254T 8206 8234 8238T 8232W 8234T 8238 8240W 8253W 8244T 8240 8224T 8214T 8252 8254W 8202T 8236W 8242T 8232T 8253T 8214 8228T 8224W 8210T 8236T 8252W 8202W 8238W 8240T 8216T 8242W
14216-662EA 474.71 USD InStock
14216-662 14216-664 14216-740 14216-586 14216-622 14216-666 14216-588 14216-668 14216-624 14216-660 14216-736 14216-658 14216-614 14216-738 14216-574 14216-650 14216-576 14216-578 14216-610 14216-612 14216-690 14216-692 14216-648 14216-724 14216-726 14216-728 14216-684 14216-562 14216-686 14216-564 14216-600 14216-688 14216-566 14216-646 14216-602 14216-682 14216-714 14216-636 14216-716 14216-638 14216-552 14216-672 14216-598 14216-554 14216-674 14216-712 14216-634 14216-590 14216-550 14216-670 14216-626 14216-702
Electronic Balance Calibration Weights, ANSI/ASTM Class 2 Weight, Troemner
Weights Calibration Weights
Precision-crafted of stainless steel, with a highly polished finish
Weights 5kg and below supplied in a rugged, precision-fit polypropylene case with handling glove.
Standard weights are supplied with a Statement of Accuracy. Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking. Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straight cylinders. All are manufactured with adjusting cavities.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty