Would you like to log in again?
Stat Zap 2102 Metalized Static Shielding Bags, 3.1 mil, SECO
Supplier: SECO
Stat Zap
212Z-68 212-810 212-1212 212Z-912 212-812 212-1218 212-810 212Z-1518 212-1818 212-1518 212Z-1218 212Z-1012 212-1416 212Z-1215 212-1012 212Z-1818 212-912 212-35 212-1215 212Z-1212 212Z-1416 212-46 212Z-1016 212-68
14218-084PK 68.3 USD InStock
14218-084 14218-086 14218-140 14218-074 14218-092 14218-070 14218-102 14218-146 14218-116 14218-138 14218-104 14218-148 14218-132 14218-088 58605-918 14218-098 14218-142 14218-076 14218-134 14218-100 14218-144 14218-128 14218-106 14218-108
Stat Zap 2102 Metalized Static Shielding Bags, 3.1 mil, SECO
Bags Sample Bags
Transparent, metalized static shielding creates a Faraday Cage effect with E.M.I.
  • R.F.I. protection provides a static-safe environment for sensitive electronic devices
Sandwiching the metal layer between polyester and polyethylene enhances durability of these bags. Bags are hot stamped with a yellow ESD warning symbol and date coded for QC traceability.
Certifications: Tested to meet or exceed the electrical and physical requirements of MIL-B-81705C Type II and EIA 541.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty