Would you like to log in again?
Orion™, All-in-One™ pH Buffers Kit, Thermo Scientific
Supplier: Thermo Scientific
910199 810199 810199
97034-758EA 174.22 USD InStock
CA97034-758 97034-758 14218-368
Orion™, All-in-One™ pH Buffers Kit, Thermo Scientific
Buffers pH Calibration Buffers
This pH buffer kit has colour coded buffers, storage solution and storage bottles.
They are manufactured under ISO 9000 quality standards. The kit includes 1 pint (475 ml) bottle each of pH 4.00 buffer, pH 7.00 buffer, pH 10.01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.
pH buffers with temperature tables on the labels are avilable for accurate calibrations and NIST™ traceability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty