Would you like to log in again?
Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
081045
14218-404EA 262.28 USD InStock
14218-404
Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes, Thermo Scientific
Electrode Holders
Provide additional protection and weight with Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes.
For Use With
Conductivity Cells: 013005MD, 013010MD, 0130020MD, 013025MD, 013005A, 013010A, and 013005D
Dissolved Oxygen Probes: 080510MD, 0830005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, and 083005D
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty