Would you like to log in again?
Monoject™ ANGEL WING® Blood Collection Sets, Covidien
Supplier: Covidien
Monoject™
8881225174 8881225299 8881225707 8881225190 8881225182 8881225265 8881225281 8881225208 8881225273 8881225315 8881225307 8881225390
84009-740CS 310.98 USD InStock
84009-740 14219-052 37001-292 14219-050 14219-208 14219-056 14219-210 82013-182 14219-054 14219-064 14219-214 14219-212
Monoject™ ANGEL WING® Blood Collection Sets, Covidien
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
All sets feature easy-to-handle textured wings and one-handed activation of the safety mechanism
The blood collection sets with attached blood tube/needle holders feature a unique self-sheathing needle system and superior needle sharpness.
All sets come with a 1.9cm (3/4") needle. Sets with a female luer connector are compatible with all blood drawing equipment. A luer adapter is also available for multi-sample collection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty