Would you like to log in again?
VWR® Autoclavable Bags, 1.5 mil
Supplier: VWR
VWR®
14220-022 14220-020 14220-016 14220-008 14220-006 14220-018 14220-002 14220-004 14220-010 14220-012 14220-014 14220-000 14220-024
14220-008CS 599.1 USD InStock
14220-008 14220-012 14220-022 14220-000 14220-014 14220-002 14220-024 14220-016 14220-004 14220-018 14220-006 14220-010 14220-020
VWR® Autoclavable Bags, 1.5 mil
Bags Waste Disposal Bags
Autoclave bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
  • Bags are available clear or red with proper biohazard warnings, or clear without label
  • Rated for use at 135 °C (275 °F)
High-strength, highly temperature-resistant polypropylene construction allows perfect adaptability to standard laboratory disposal routines, including steam autoclaving.
Ordering information: Twist ties included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty