Would you like to log in again?
VWR® Autoclavable Bags, Nonhazardous Waste
Supplier: VWR
VWR®
14220-038 14220-044 14220-040 14220-036 14220-042
14220-040CS 186.72 USD InStock
14220-040 14220-042 14220-044 14220-036 14220-038
VWR® Autoclavable Bags, Nonhazardous Waste
Bags Waste Disposal Bags
Intended for use in autoclaving nonhazardous waste and constructed of high-integrity polypropylene, these bags can withstand autoclaving temperatures up to 135 °C (275 °F).
  • Superior tear resistance
  • Welded seals
Heavy walls provide superior tear resistance and welded seals minimize risk of leakage. Bags meet the strength requirements of the 165g dart drop standard performed per ASTM D1709-98. All bags are clear.
Certifications: Meets the 165 g dart drop standard (ASTM D1709-98).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty