Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® High Strength Autoclavable Infectious Waste Bag
Supplier: VWR
VWR®
11215-816 14230-910 14220-098 14220-096 14220-092 14220-094 14220-104 14230-912 14220-100 14220-102 14220-090
14220-092CS 832.63 USD InStock
14220-092 14220-094 14220-096 14220-098 14230-912 14220-100 14230-910 14220-102 14220-104 11215-816 14220-090
VWR® High Strength Autoclavable Infectious Waste Bag
Bags Waste Disposal Bags
These 2 mil polypropylene bags are engineered to meet the most demanding regulatory specifications worldwide.
  • Designed to handle dangerous materials
  • High-strength
  • Autoclavable
They exceed the 480 gram tear test as defined by ASTM D1922 in both length (MD) and width (TD) directions.
These high strength bags are ideal for hazardous waste collection and decontamination in an autoclave. Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory.
These bags are red and are available printed with a black biohazard symbol or plain. The printed bags also read 'Infectious Waste' and feature a temperature indicator patch. All bags are autoclavable to 135 °C (275 °F).
Certifications: Bags meet all state requirements for hazardous waste handling, including Florida, Pennsylvania and California Department of Health codes. ASTM D1922 for tear resistance greater than 480 grams in both width and height of bag.
Ordering information: Bags are supplied with 12.7 cm (5) twist ties.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty