Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Incineration Bags
Supplier: Avantor
VWR®
14220-112 14220-108 14220-114 14220-110 14220-106
14220-108CS 167.42 USD InStock
14220-108 14220-110 14220-112 14220-114 14220-106
VWR® Incineration Bags
Bags Waste Disposal Bags
Constructed of durable polypropylene that meets the 165 g dart drop standard performed per ASTM D1709-98 and the 480 g tear standard performed per ASTM D1299-94A.
  • Durable polypropylene construction
  • Not autoclavable
  • Marked with biohazard symbol
Red bags feature the universal biohazard symbol in black.
Note: Autoclaving not recommended. Bags will melt at 98.9 °C (210 °F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty