Would you like to log in again?
Borosilicate Glass VOC Vials, DWK Life Sciences
Supplier: DWK Life Sciences
W227358
14221-532CS 244.13 USD InStock
14221-532
Borosilicate Glass VOC Vials, DWK Life Sciences
Vials
These autosampler vials with open top caps are ideal for the analysis of volatile organic compounds.
  • Thick septa (3.2 mm)
  • Heavy-working
  • Long time injection
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty