Would you like to log in again?
WHEATON® Open Top Sample Vials In Lab File, DWK Life Sciences
Supplier: DWK Life Sciences
W227356
14221-606CS 240.93 USD InStock
14221-606
WHEATON® Open Top Sample Vials In Lab File, DWK Life Sciences
Vials
These vials are manufactured from high-quality glass that meets ASTM standards.
  • Amber finish
  • Septa are bonded to the cap providing a permanent connection
Glass meets USP Type 1 standards to protect light sensitive samples and maintain purity of contents. The white polypropylene open top screw cap has a specially designed septum of 5 mils of Teflon facing 0.120 mils of silicone. The polypropylene is glass-filled to provide extra chemical and heat resistance compared to standard polypropylene.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty