Would you like to log in again?
TWD Scientific Weighing Funnels
Supplier: TradeWinds (TWD)
TWD Scientific
DPWF-PP2-M GWF-BS3-L ASWFXS DPWF-PP1-S ASWF2M GWF-ASSORT GWF-BS1-S ASWF1S ASWF4XL DPWF-ASSORT GWF-BS2-M DPWF-PP4-XL ASWF3L GWF-BS-XS ASWF-ASSORT ASWF5XXL DPWF-PP3-L GWF-BS4-XL DPWF-PP-XS
77172-148PK 176.33 USD InStock
77172-148 14221-818 14221-808 14221-810 14221-800 14221-822 14221-812 14221-824 14221-802 14221-814 14221-804 14221-826 14221-816 14221-828 14221-806 77172-154 77172-152 77172-150 14221-820
TWD Scientific Weighing Funnels
Boats Weighing Boats
The TWD Scientific (formerly Tradewinds Direct) family of weighing funnels (weighing boats) all feature a flat bottom for sitting on balances and a pouring neck that fits to volumetric flasks, micro-tubes, or any narrow neck apparatus for spill-free transfer.
  • Available in antistatic polypropylene, disposable polypropylene, and borosilicate glass
  • Residual samples can be rinsed to guarantee 100% transfer
  • Five different sizes of funnels fit to various flask sizes
  • X-Small funnels fit 96-well mini-tubes and deep well plates
Suitable for safe handling of hazardous materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty