Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Autoclavable Biohazard Bags and Containers
Supplier: VWR
VWR®
14220-070 1412-1924VWR 14220-066 14221-996 1412-1419VWR 14221-994 1412-3748VWR 14220-026 11215-904 14220-054 11215-680 14220-032 1412-2436VWR 14221-164 1412-0812VWR 14220-028 14220-030 14220-048 14220-034 14221-156 14220-052 14220-068 14220-050 14220-046
89129-470CS 253.06 USD InStock
89129-470 14220-034 14220-066 14220-068 14220-046 89129-474 14220-026 14220-048 11215-904 89129-472 14220-028 14220-070 14220-050 14220-030 14221-156 14220-052 14220-054 14220-032 89129-468 14221-164 89130-796 14221-994 14221-996 11215-680
VWR® Autoclavable Biohazard Bags and Containers
Bags Waste Disposal Bags
Double-thick bags are ideal for sterilizing animal cages and other objects with sharp corners.
  • High-integrity polypropylene
  • Available with temperature indicator
  • Resists punctures, tears, and leaks
Ideal for processing biohazardous materials that require autoclaving prior to disposal.
Twist-tie closure allows bag to expand and vent steam during cooling. Bags resist punctures, tears, and leaks and meet the 165g dart drop standard performed as required by ASTM D1709-98. Each bag is imprinted with the universal biohazard symbol.
Indicator bags have the words 'Contents Have Been Autoclaved' printed in a chemically active ink. Before autoclaving, the ink is white. After autoclaving is complete, it darkens, providing instant proof that biohazardous waste has been autoclaved.
Biohazard bag containers are made of rigid, high-density polyethylene for durability and leak-free performance. Snap-on, snap-off lid provides an extra measure of safety in the lab. White with universal biohazard waste symbol imprinted in black on an orange background. Containers are not autoclavable.
Certifications: Meets the 165 g dart drop standard (ASTM D1709-98).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty