Would you like to log in again?
TAPE TINTED BLUE 2IN
Supplier: ULTRATAPE INDUSTRIES
Conductive Grid Tape^ Conductive grids embedded between two strong polypropylene layers. Approximately 100V maximum upon removal from most surfaces. Static-shielding. No residues. Cardboard core. Silicone-free. Length: 33m. Width: 5.1cm. Color: tinted blue.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TAPE TINTED BLUE 2IN 14222-232 1112TB200P3D Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty