Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SCOTT® Kitchen Roll Towels, KIMBERLY-CLARK PROFESSIONAL®

Supplier: Kimberly-Clark

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SCOTT®
41482 41482
14222-321CS 100.78 USD InStock
CA14222-321 14222-321
SCOTT® Kitchen Roll Towels, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Paper Towels
SCOTT® Brand consistently delivers superior value by offering the optimum balance of product quality and affordability.

  • More sheets on a roll to last longer
  • Smaller case for easier handling and storage
  • 4.9" roll Ø

Made with ABSORBENCY POCKETS™ for a soft and absorbent performance to handle tough spills and clean-ups. Meets EPA standards with a minimum of 40 % post-consumer waste content; 40% post-consumer waste; 80% recycled fiber.

Certifications: FSC & EcoLogo Certified.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty