Would you like to log in again?
Orion™ Surfactant Ion Selective Electrode, Thermo Scientific
Supplier: Thermo Scientific
9342BN
14223-118EA 2039.94 USD InStock
14223-118
Orion™ Surfactant Ion Selective Electrode, Thermo Scientific
Electrodes ISE Electrodes
This electrode indicates endpoint in titration of anionic or cationic surfactants in solutions.
Designed for use on Orion 960 Autochemistry system.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty