Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Orion™ Automatic Stirrer Probe and Paddle, Thermo Scientific
Supplier: Thermo Scientific
96019 96021
14223-126EA 463.91 USD InStock
14223-126 14223-214
Orion™ Automatic Stirrer Probe and Paddle, Thermo Scientific
Stirrers Stirring Tools
Use the Orion™ Star Stirrer Probe with Orion™ benchtop meters to stir solutions for better accuracy without the difficulty of using a stir plate and bar.
Use the Orion™ Star Stirrer Probe with Orion™ Versa Star™ meters, Orion™ Star A210 series benchtop meters, and legacy Orion™ Star series benchtop meters to stir standards and samples for better measurement accuracy without the difficulty of separate stir plate and bars. The Orion™ Star Stirrer Probe is powered and controlled by the meter with five optimized stirring speeds. The device fits in the electrode stand for easy movement in and out of samples and can be cleaned simply by rinsing. Paddles can be replaced to extend the life of Orion™ Star Stirrer Probes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty