Would you like to log in again?
Environmental Quality Meter, Sper Scientific
Supplier: Sper Scientific
850071
14223-290EA 360.92 USD InStock
14223-290
Environmental Quality Meter, Sper Scientific
Anemometers
Using interchangeable probes (sold separately) this versatile environmental meter reads airspeed, air temperature, RH, light and thermocouple and IR temperature
You can order probes for only the parameters currently required and add additional probes in the future. Intelligent probes electronically retain calibration information. No recalibration is required when changing probes. The display simultaneously reads temperature in user selectable ºC and ºF and all measurements are automatically temperature compensated. The RS232 port enables communication with a computer. The environmental quality meter also features automatic shut off, hold and min/max, has a fold out easel back and a tripod screw.
Ordering information: Comes with a 9V battery and a hard-shell foam-lined carrying case with room for one or two probes.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty