Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
UVP ChromaDoc-It™ TLC Imaging System, Analytik Jena
Supplier: Analytik Jena
ChromaDoc-It™
97-0109-02 97-0109-01
14223-344EA 6856.62 USD InStock
14223-344 14223-346
UVP ChromaDoc-It™ TLC Imaging System, Analytik Jena
Photodocumentation Systems
The ChromaDoc-It™ system is ideal for simple, accurate and repeatable quantitative determination of TLC plates. System includes a digital color camera, a Chromato-Vue® cabinet, and image capture software for brilliant color TLC images.
The image capture software is compatible with Windows® 98, ME, and 2000 operating systems with a USB interface
The optional Doc-It® 1D analysis software loads on a separate computer for analysis of gels, plates, and membranes. Features include automatic lane and band detection, quantitative analysis of lanes and bands, multiple MW calibration, background correction, image enhancement, and reporting capabilities.
The digital camera features 5.0 megapixels, 640x480 to 2592x1944 resolution, 4x optical zoom lens, TLC filters, and a diopter. The cabinet features built-in overhead 254/365nm UV and white light with 15W shortwave and longwave UV tubes positioned on two sides of the cabinet for uniform illumination. Cabinet dimensions: 50.2Wx42Dx40Hcm (193/4x161/2x153/4").
The capture software controls camera functions including live image preview, focus and exposure settings, and image saturation warnings. It also features enhancement and annotation, detailed and configurable reporting, and history tracking.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty