Would you like to log in again?
Stainless Steel Cleaner, Steris
Supplier: Steris
NM410 NM410
14223-538EA 53.88 USD InStock
14223-538 CA14223-538
Stainless Steel Cleaner, Steris
Cleaning Agents Detergents and Surface Cleansers
Cleans and protects equipment with stainless steel, Formica® laminate, chrome, aluminum, and plastic finishes
Contains no fluorocarbons or ozone-depleting chemicals. Flammable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty