Would you like to log in again?
Orion™ AQUAfast® II C.O.D. Colorimeter, Thermo Scientific
Supplier: Thermo Scientific
ORION® AQUAfast®
CODS01 CODL00 CODS01 CODS10 CODHP0 CODS10
14223-554EA 118.99 USD InStock
14223-554 14232-926 CA22000-602 CA14232-926 CA14223-554 CA22000-600
Orion™ AQUAfast® II C.O.D. Colorimeter, Thermo Scientific
Environmental Test Kits
Microprocessor-based colorimeter includes a three-button user interface for error-free readings, a large LCD display, and a rugged carrying case. Unit is battery operated with a 9V battery making it ideal for field or lab use. It features a low range wavelength of 430nm and a medium/high range of 610nm. Cuvettes are not supplied with colorimeter.
The C.O.D test kits include 25 cuvettes with caps, measuring 16x92mm
The C.O.D. thermoreactor fits twenty-five 16mm vials and features variable time settings. It has temperature settings of 70°C (158°F), 100°C (212°F), 120°C (248°F), and 150°C (302°F) with an accuracy of ±0.5°C.
Kits have an accuracy of ±3.5%. The C.O.D. standards have an accuracy of ±0.5% at the time of bottling and a shelf life of two years. Fully prepared test reagents provide convenience and safety for the user.
Certifications: Reagents comply with EPA guidelines for COD testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty