Would you like to log in again?
Orion™ Powder Chemistry Packs, Thermo Scientific
Supplier: Thermo Scientific
Orion™
AC4P42 AC4P46 AC4P72 AC4P78 AC4P60 AC4P82 AC4P29 AC4P95 AC4P12 AC4P12 AC4P72 AC4P79 AC4P71 AC4P79 AC4P82 AC4P55 AC4P46 AC4P55 AC4P95 AC4P71 AC4P29 AC4P78 AC4P42 AC4P60
14223-562PK 47.95 USD InStock
14223-562 CA14223-560 14223-574 CA14223-572 14223-564 CA14223-562 14223-576 CA14223-574 14223-570 14223-560 14223-572 CA14223-570 CA14223-568 CA14223-558 CA14223-564 14223-566 14223-578 14223-556 CA14223-576 CA14223-566 14223-568 CA14223-578 14223-558 CA14223-556
Orion™ Powder Chemistry Packs, Thermo Scientific
Environmental Test Kits
Orion™ AQUAfast Powder Pack Reagents perform colorimetric measurements of common water parameters with powders that dissolve quickly and mix easily.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast Colorimeters
Orion™ AQUAfast Powder Pack Reagents are designed for use with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Powder pack chemistries dissolve quickly in solution, are easy to mix, and do not require a stir rod or crushing. Each powder pack is notched for simplified opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica, and sulfate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty