Would you like to log in again?
Common Point Grounding Cord, STATICO
Supplier: Statico
S1030NRF
14223-734EA 16.4 USD InStock
14223-734
Common Point Grounding Cord, STATICO
Grounders Grounding Wires
This cord is a heavy duty 15' long low-profile wrist strap/table mat Common Point Grounding Cord (CPG) with two banana plug receptacles
This product can be snapped on the mat. It has no resistor and provides a direct pass to the ground.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty