Would you like to log in again?
Double Stack Connector, Statico
Supplier: Statico
S1092-38MFB
14223-736EA 25.63 USD InStock
14223-736
Double Stack Connector, Statico
Grounders Grounding Wires
Molded male/female double-stack snap with banana jack and caps. Provides a connecting point for:
  • Wrist grounding strap via banana jack
  • Snaps-on table mat either via female or male 3/8" (10 mm) socket
Either one of the unused snaps can be covered by a plastic cap, to prevent shock due to exposure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty