Would you like to log in again?
Disposable Heel Grounder, STATICO
Supplier: Statico
S1100V
14223-828PK 164.21 USD InStock
14223-828
Disposable Heel Grounder, STATICO
Grounders Heel and Toe Grounders
The S1100 Series is an easy to use disposable heel grounder that dissipates the static charges effectively, while offering convenience of use
It is designed for use by visitors and mobile personnel at the static sensitive work areas where protection against ESD (Electro Static Discharge) is required. Use of two-foot grounders is required for dependable protection and to meet most ESD control standards.
Certifications: Meets ANSI/ESD S1.1-2013 & ANSI/ESD S20.20-2014.
Ordering information: Heel grounders are packaged 100 per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty