Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HyClone™ SFM4MAb media, Cytiva
Supplier: HyClone products (Cytiva)
HyClone™
SH30535.02 SH30513.02 SH30535.02 SH30513.02
14224-300EA 97.17 USD InStock
CA76462-528 14224-300 CA14224-300 76462-528
HyClone™ SFM4MAb media, Cytiva
Cell Culture Media
HyClone™ SFM4MAb media was developed through the Metabolic Pathway Design approach to increase process yields for manufacturing of human and humanised recombinant antibodies for therapeutic use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty